Learn More
Aviva Systems Biology Corp C18orf10 Antibody - middle region (ARP55267_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55267_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C18orf10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C18orf10. Alternate names: DKFZP586M1523, HMFN0601, HsT3006, L17, PGs2, C18orf10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61452