Learn More
Cayman Chemical C19 CeramIded18 119 0 10mg
SDP
Catalog No. 502933216
Encompass_Preferred
A naturally occurring ceramide; elevated in adult and decreased in juvenile whole rat brain extracts by 114 and 37%, respectively, following chronic ethanol exposure; increased in mouse hearts following administration of angiotensin II
Catalog No. 50-293-3216 Supplier Cayman Chemical Supplier No. 2439510 MG
A naturally occurring ceramide; elevated in adult and decreased in juvenile whole rat brain extracts by 114 and 37%, respectively, following chronic ethanol exposure; increased in mouse hearts following administration of angiotensin II