Learn More
Aviva Systems Biology Corp C19orf21 Antibody - C-terminal region (ARP53237_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53237_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C19orf21. Alternate names: DKFZp686H18209, C19orf21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53705