Learn More
Aviva Systems Biology Corp TEX45 Antibody - middle region (ARP55899_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55899_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf45. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C19orf45. Alternate names: FLJ35784, FLJ56642ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-286-093