Learn More
Aviva Systems Biology Corp C19orf46 Antibody - N-terminal region (ARP44895_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C19orf46. Alternate names: FLJ36445, SYNE4, C19orf46ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-58931