Learn More
Aviva Systems Biology Corp C19orf47 Antibody - middle region (ARP48452_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48452_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf47. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C19orf47. Alternate names: DKFZp686P05129, FLJ36888ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59831