Learn More
Aviva Systems Biology Corp C19orf47 Antibody - C-terminal region (ARP48453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf47. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C19orf47. Alternate names: DKFZp686P05129, FLJ36888ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59832