Learn More
Aviva Systems Biology Corp PROSER3 Antibody - middle region (ARP51861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf55. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C19orf55. Alternate names: DKFZp434C2231, FLJ30657, MGC131952ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62895