Learn More
Aviva Systems Biology Corp C19orf56 Antibody - N-terminal region (ARP47218_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf56. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C19orf56. Alternate names: PTD008, C19orf56ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59488