Learn More
Aviva Systems Biology Corp C19ORF6 Antibody - middle region (ARP34137_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34137_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C19ORF6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human C19ORF6. Alternate names: MBRL, ASBABP1, R32184_3, MEMBRALIN, C19ORF6ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-52941