Learn More
Aviva Systems Biology Corp C19orf62 Antibody - N-terminal region (ARP54986_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54986_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C19orf62. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C19orf62. Alternate names: FLJ20571, HSPC142, NBA1, MERIT40, C19orf62ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53746