Learn More
Aviva Systems Biology Corp C1orf102 Antibody - N-terminal region (ARP53512_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53512_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf102. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C1orf102. Alternate names: MGC26685, NOR1, OSCP1, C1orf102ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61040