Learn More
Aviva Systems Biology Corp C1orf103 Antibody - N-terminal region (ARP56173_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56173_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf103. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C1orf103. Alternate names: FLJ11269, RIF1, RP11-96K19.1, C1orf103ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61746