Learn More
Aviva Systems Biology Corp CCDC190 Antibody - middle region (ARP55776_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55776_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf110. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf110. Alternate names: FLJ41579, MGC48998, RP11-331H2.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61607