Learn More
Aviva Systems Biology Corp SPATA46 Antibody - N-terminal region (ARP48011_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf111. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C1orf111. Alternate names: HSD20, RP11-565P22.3ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit
Catalog No. 50-285-338