Learn More
Aviva Systems Biology Corp SPATA46 Antibody - middle region (ARP48010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf111. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf111. Alternate names: HSD20, RP11-565P22.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-54066