Learn More
Aviva Systems Biology Corp C1orf116 Antibody - middle region (ARP41630_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf116. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf116. Alternate names: DKFZp666H2010, FLJ36507, MGC2742, MGC4309, SARGApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57928