Learn More
Aviva Systems Biology Corp C1orf122 Antibody - N-terminal region (ARP53467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf122. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human C1orf122. Alternate names: ALAESM, FLJ23476, FLJ45459, C1orf122ApplicationWB. Species reactivity: Human
Catalog No. 50-014-63034