Learn More
Aviva Systems Biology Corp C1orf159 Antibody - C-terminal region (ARP47374_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47374_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf159. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C1orf159. Alternate names: RP11-465B22.4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59528