Learn More
Aviva Systems Biology Corp C1orf174 Antibody - middle region (ARP55999_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55999_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf174. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf174. Alternate names: RP13-531C17.2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61689