Learn More
Aviva Systems Biology Corp LEXM Antibody - middle region (ARP53062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf177. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf177. Alternate names: FLJ40201ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60908