Learn More
Aviva Systems Biology Corp INKA2 Antibody - middle region (ARP57804_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57804_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf183. It was validated on Western Blot. C1orf183,chromosome 1 open reading frame 183,FLJ31105; C1orf183,hypothetical protein LOC55924,hypothetical protein LOC55924The immunogen is a synthetic peptide directed towards the middle region of human C1orf183. Alternate names: FLJ31105, C1orf183ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-286-386