Learn More
Aviva Systems Biology Corp C1orf216 Antibody - N-terminal region (ARP52922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf216. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C1orf216. Alternate names: FLJ38984ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-785