Learn More
Aviva Systems Biology Corp C1ORF25 Antibody - N-terminal region (ARP30109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1ORF25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C1ORF25. Alternate names: TRM1L, MST070, C1orf25, MSTP070, bG120K12.3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54129