Learn More
Aviva Systems Biology Corp C1orf43 Antibody - middle region (ARP54537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf43. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf43. Alternate names: DKFZp586G1722, HSPC012, MGC111001, NICE-3, NS5ATP4, S863-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54006