Learn More
Aviva Systems Biology Corp C1orf74 Antibody - N-terminal region (ARP52998_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52998_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf74. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C1orf74. Alternate names: FLJ25078ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60892