Learn More
Aviva Systems Biology Corp STUM Antibody - middle region (ARP44446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf95. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf95. Alternate names: DKFZp761P211, RP11-9C4.1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-58811