Learn More
Aviva Systems Biology Corp C1orf96 Antibody - middle region (ARP52835_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52835_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1orf96. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1orf96. Alternate names: FLJ37296, FLJ41471, RP4-803J11.3, C1orf96ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-772