Learn More
Complement Technology Inc C1q Protein, Catalog Number A099, 1000 μg/vial, 1.0 mg/mL, Frozen liquid, > 98% purity, 410,000 Da, 10 mM HEPES, 300 mM NaCl, pH 7.2, Negative for HBsAg, STS, HIV
Supplier:  Complement Technology Inc A099
Encompass
C1q Protein, Catalog Number A099, 1000 μg/vial, 1.0 mg/mL, Frozen liquid, > 98% purity, 410,000 Da, 10 mM HEPES, 300 mM NaCl, pH 7.2, Negative for HBsAg, STS, HIV
Catalog No. NC2213077