Learn More
Diagnostic Biosystems C1Q-FITC C1q Complement-FITC
Supplier:  Diagnostic Biosystems F010
Encompass_Preferred
This antibody reacts with the human C1q complement. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7583