Learn More
MyBioSource Complement Component C1q Native Protein, 0.2 mg
SDP
Catalog No. NC1913445
Encompass_Preferred
Human Complement Component C1q produced in Human plasma having a molecular mass of 410 kDa.
Catalog No. NC1913445 Supplier MyBioSource Supplier No. MBS1473050.2 MG
Human Complement Component C1q produced in Human plasma having a molecular mass of 410 kDa.