Learn More
MyBioSource Native Protein, Complement Component C1q
SDP
Catalog No. NC2295426
Encompass_Preferred
The product is human complement component C1q protein. It is a 460 amino acid protein that is part of the complement system and helps activate the classical pathway. The protein is purified from human plasma and is supplied lyophilized from a 0.2 μm filtered solution in PBS. The molecular weight is approximately 460 kDa. This research-grade reagent is suitable for in vitro experiments studying complement activation and inflammation.
Catalog No. NC2295426 Supplier MyBioSource Supplier No. MBS1473051 MG
The product is human complement component C1q protein. It is a 460 amino acid protein that is part of the complement system and helps activate the classical pathway. The protein is purified from human plasma and is supplied lyophilized from a 0.2 μm filtered solution in PBS. The molecular weight is approximately 460 kDa. This research-grade reagent is suitable for in vitro experiments studying complement activation and inflammation.