Learn More
Aviva Systems Biology Corp C1QA Antibody - N-terminal region (ARP60064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1QA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C1QA. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-933