Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C1QA Mouse His-SUM 500ug
SDP

Catalog No. 5000195942
Encompass_Preferred

Recombinant mouse C1QA (mature residues E23-A245, accession P98086) expressed in E. coli with an N-terminal 6xHis-SUMO tag. Supplied lyophilized and formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0); purity >90% by reducing SDS-PAGE and apparent molecular weight ~42 kDa. Intended for research use only.

  • Contains full-length mature C1QA sequence (E23-A245).
  • N-terminal 6xHis and SUMO tags for affinity purification and improved solubility.
  • Produced in Escherichia coli and supplied lyophilized for stability.
  • Purity greater than 90% by reducing SDS-PAGE; ~42 kDa apparent molecular weight.
  • Formulated in Tris-EDTA with trehalose to protect during storage and freeze-thaw.
  • Stable at -20°C for long-term storage; reconstitute to at least 100 μg/mL for handling.

Catalog No. 50-001-95942 Supplier Medchemexpress LLC Supplier No. HYP72108500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse C1QA (mature residues E23-A245, accession P98086) expressed in E. coli with an N-terminal 6xHis-SUMO tag. Supplied lyophilized and formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0); purity >90% by reducing SDS-PAGE and apparent molecular weight ~42 kDa. Intended for research use only.

  • Contains full-length mature C1QA sequence (E23-A245).
  • N-terminal 6xHis and SUMO tags for affinity purification and improved solubility.
  • Produced in Escherichia coli and supplied lyophilized for stability.
  • Purity greater than 90% by reducing SDS-PAGE; ~42 kDa apparent molecular weight.
  • Formulated in Tris-EDTA with trehalose to protect during storage and freeze-thaw.
  • Stable at -20°C for long-term storage; reconstitute to at least 100 μg/mL for handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.