Learn More
Aviva Systems Biology Corp C1QB Antibody - middle region (ARP44296_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44296_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1QB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1QB. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58774