Learn More
ACROBiosystems Recombinant Protein;Human C1q R1 / CD93 Protein;HEK293;50UG;C11,CD93,CDw93,C1QR1,MXRA4,C1qR,C1qR(P),C1qRP,ECSM3,C1q R1
Catalog No. 501658999
Encompass_Preferred
Recombinant Protein;Human C1q R1 / CD93 Protein;50UG;Human C1q R1, His Tag (C11-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Lys 580 (Accession # AAH28075).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8999 Supplier ACROBiosystems Supplier No. C11H522850UG
Recombinant Protein;Human C1q R1 / CD93 Protein;50UG;Human C1q R1, His Tag (C11-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Lys 580 (Accession # AAH28075).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.