Learn More
Aviva Systems Biology Corp C1QTNF7 Antibody - middle region (ARP52426_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52426_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C1QTNF7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C1QTNF7. Alternate names: CTRP7, ZACRP7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60739