Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C1s-A subcomponent protein | >85.0% | 10 UG
SDP

Catalog No. 5000293004
Encompass_Preferred

Recombinant mouse C1s-A subcomponent serine protease expressed in HEK293 cells and supplied as a filtered PBS solution with a C-terminal His tag. Intended for research use in studies of classical complement pathway activation; follow handling and storage recommendations provided in the product documentation.

  • Species: Mouse.
  • Expression system: HEK293 cells.
  • Tag: C-terminal 10xHis.
  • Formulation: 0.22 μm filtered PBS, pH 7.4.
  • Purity: >85.0% by reducing SDS-PAGE.
  • Endotoxin: <1 EU/μg (LAL method).
  • Unit size: 10 μg.
  • Storage: store at -80°C; avoid repeated freeze-thaw.
  • Shipping: shipped on dry ice.

Catalog No. 50-002-93004 Supplier Medchemexpress LLC Supplier No. HYP7546310UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse C1s-A subcomponent serine protease expressed in HEK293 cells and supplied as a filtered PBS solution with a C-terminal His tag. Intended for research use in studies of classical complement pathway activation; follow handling and storage recommendations provided in the product documentation.

  • Species: Mouse.
  • Expression system: HEK293 cells.
  • Tag: C-terminal 10xHis.
  • Formulation: 0.22 μm filtered PBS, pH 7.4.
  • Purity: >85.0% by reducing SDS-PAGE.
  • Endotoxin: <1 EU/μg (LAL method).
  • Unit size: 10 μg.
  • Storage: store at -80°C; avoid repeated freeze-thaw.
  • Shipping: shipped on dry ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.