Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C1s-A subcomponent protein, mouse | >85.0% | 50 UG
SDP

Catalog No. 5000291424
Encompass_Preferred

Recombinant mouse C1s-A subcomponent is a His-tagged serine protease produced in HEK293 mammalian cells and supplied as a 0.22 μm filtered PBS solution for research use. It is provided in microgram-scale vials intended for in vitro studies of the classical complement pathway.

  • His-tagged for affinity purification and detection.
  • Expressed in HEK293 mammalian cells to support native folding and glycosylation.
  • Supplied in PBS, pH 7.4, 0.22 μm filtered solution for immediate use.
  • Packaged as a 50 μg vial suitable for assay development and biochemical studies.
  • Purity >85.0% by reducing SDS-PAGE for reliable performance.
  • Stable when stored at -80°C and shipped on dry ice; avoid repeated freeze-thaw cycles.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-91424 Supplier Medchemexpress LLC Supplier No. HYP7546350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse C1s-A subcomponent is a His-tagged serine protease produced in HEK293 mammalian cells and supplied as a 0.22 μm filtered PBS solution for research use. It is provided in microgram-scale vials intended for in vitro studies of the classical complement pathway.

  • His-tagged for affinity purification and detection.
  • Expressed in HEK293 mammalian cells to support native folding and glycosylation.
  • Supplied in PBS, pH 7.4, 0.22 μm filtered solution for immediate use.
  • Packaged as a 50 μg vial suitable for assay development and biochemical studies.
  • Purity >85.0% by reducing SDS-PAGE for reliable performance.
  • Stable when stored at -80°C and shipped on dry ice; avoid repeated freeze-thaw cycles.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.