Learn More
Hycult Biotech Inc C1S/C1-INH Human ELISA Kit, 2 x 96 detections, 1.56 to 100 ng/ml standard range, 100 μl/well working volume, storage at 4 °C
SDP
Supplier:  Hycult Biotech Inc HK39902
Encompass
C1S/C1-INH Human ELISA Kit, 2 x 96 detections, 1.56 to 100 ng/ml standard range, 100 μl/well working volume, storage at 4 °C
Catalog No. NC2544149