Learn More
Complement Technology Inc C1s Enzyme, 250 ug
Catalog No. NC1749965
Encompass
C1s enzyme is the activated form of C1s proenzyme. C1s is a subunit of the C1complex which is the first complement component in the classical pathway ofcomplement. C1s proenzyme is an inactive zymogen until C1 is activated. C1sproenzyme is activated when C1 binds to and is activated by antibodies bound to antigens(immune complexes) yielding C1r enzyme, the first protease that initiates the cascade.
Catalog No. NC1749965 Supplier Complement Technology Inc Supplier No. A104
C1s enzyme is the activated form of C1s proenzyme. C1s is a subunit of the C1complex which is the first complement component in the classical pathway ofcomplement. C1s proenzyme is an inactive zymogen until C1 is activated. C1sproenzyme is activated when C1 binds to and is activated by antibodies bound to antigens(immune complexes) yielding C1r enzyme, the first protease that initiates the cascade.