Learn More
Elemental Scientific Inc C2-140228RH04-500 STANDARD
SDP
Catalog No. NC0656881
Encompass_Preferred
Custom Standard, APS WRF/PVNGS IC3000 High Anion B Second Source, 500 ml, 150 mg/L- PO4; 300m mg/L- F; 2000 mg/L- NO3 as N in H2O
Catalog No. NC0656881 Supplier Elemental Scientific Inc Supplier No. C2140228RH06500
Custom Standard, APS WRF/PVNGS IC3000 High Anion B Second Source, 500 ml, 150 mg/L- PO4; 300m mg/L- F; 2000 mg/L- NO3 as N in H2O