Learn More
Enzo Life Sciences C2 Ceramide (25 mg). CAS: 3102-57-6
SDP
Supplier:  Enzo Life Sciences BMLSL1000025
Encompass_Preferred
N-Acetylsphingosine is a biologically active, cell-permeable ceramide analog. It inhibits cell proliferation and induces monocytic differentiation of HL-60 cells and induces apoptosis. It stimulates protein phosphatase 2A4, activates MAP kinase and SAP kinase and induces PKCδ and ε translocation. Physiological levels of C2 ceramide, produced by the action of a PAF:sphingosine CoA-independent transacetylase, have been detected in HL-60 cells. Alternative name: N-Acetyl-D-erythro-sphingosine. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (>25mg/ml) or DMSO (>50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2101