Learn More
Medchemexpress LLC C2 ceramide | 3102-57-6 | MFCD00153903 | 99.9% | 341.53 | C20H39NO3 | 5 MG
SDP
Catalog No. 5000212553
Encompass_Preferred
C2 ceramide is a short-chain ceramide reagent used in cell biology to activate protein phosphatases and probe pathways such as apoptosis, autophagy, and mitochondrial function. Supplied as a small-quantity solid (e.g., 5 mg), it has molecular formula C20H39NO3 and CAS 3102-57-6; follow sealed, moisture-free storage and in-solvent cold storage recommendations for stability.
  • Activates protein phosphatase 1 (PP1), PP2A, and ceramide-activated protein phosphatase.
  • Induces cell differentiation, autophagy, and apoptosis in cellular assays.
  • Inhibits mitochondrial respiratory chain complex III for metabolic studies.
  • Available in small sizes suitable for research workflows.
  • High purity suitable for biological applications.
  • Stable when stored sealed and protected from moisture; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-12553 Supplier Medchemexpress LLC Supplier No. HY1011805MG
C2 ceramide is a short-chain ceramide reagent used in cell biology to activate protein phosphatases and probe pathways such as apoptosis, autophagy, and mitochondrial function. Supplied as a small-quantity solid (e.g., 5 mg), it has molecular formula C20H39NO3 and CAS 3102-57-6; follow sealed, moisture-free storage and in-solvent cold storage recommendations for stability.
  • Activates protein phosphatase 1 (PP1), PP2A, and ceramide-activated protein phosphatase.
  • Induces cell differentiation, autophagy, and apoptosis in cellular assays.
  • Inhibits mitochondrial respiratory chain complex III for metabolic studies.
  • Available in small sizes suitable for research workflows.
  • High purity suitable for biological applications.
  • Stable when stored sealed and protected from moisture; in solvent: -80°C (6 months), -20°C (1 month).