Learn More
Medchemexpress LLC C2 ceramide | 3102-57-6 | MFCD00153903 | 99.9% | 341.53 | C20H39NO3 | 50 MG
SDP
Catalog No. 5000213167
Encompass_Preferred
C2 Ceramide is a short-chain ceramide used as a research reagent to study ceramide signaling, protein phosphatase activation, apoptosis, autophagy, and skin barrier function. Supplied as a solid at high purity, it is intended for laboratory biochemical and cell-based assays under recommended storage conditions.
  • Activates protein phosphatases and modulates ceramide signaling.
  • Induces apoptosis and autophagy in cellular models.
  • Acts as a skin conditioning agent protecting epidermal barrier function.
  • Available in multiple pack sizes, including 50 MG, for laboratory use.
  • High purity suitable for research applications; follow recommended storage and handling.
Catalog No. 50-002-13167 Supplier Medchemexpress LLC Supplier No. HY10118050MG
C2 Ceramide is a short-chain ceramide used as a research reagent to study ceramide signaling, protein phosphatase activation, apoptosis, autophagy, and skin barrier function. Supplied as a solid at high purity, it is intended for laboratory biochemical and cell-based assays under recommended storage conditions.
  • Activates protein phosphatases and modulates ceramide signaling.
  • Induces apoptosis and autophagy in cellular models.
  • Acts as a skin conditioning agent protecting epidermal barrier function.
  • Available in multiple pack sizes, including 50 MG, for laboratory use.
  • High purity suitable for research applications; follow recommended storage and handling.