Learn More
Medchemexpress LLC C2 ceramide | 3102-57-6 | MFCD00153903 | 99.9% | 341.53 g/mol | C20H39NO3 | 1 ML
SDP
Catalog No. 5000211208
Encompass_Preferred
C2 ceramide is a short-chain, cell-permeable ceramide (N-acetylsphingosine) commonly used as a biochemical tool to probe ceramide-mediated signaling pathways, apoptosis, and phosphatase activity. Supplied as a prepared solution, it is intended for use in cell-based and biochemical assays where exogenous ceramide is required.
  • Modulates ceramide signaling and apoptosis.
  • Activates serine/threonine protein phosphatases.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity for reproducible experimental results.
  • Suitable for cell-based and biochemical assays when diluted appropriately.
Catalog No. 50-002-11208 Supplier Medchemexpress LLC Supplier No. HY10118010MM 1ML
C2 ceramide is a short-chain, cell-permeable ceramide (N-acetylsphingosine) commonly used as a biochemical tool to probe ceramide-mediated signaling pathways, apoptosis, and phosphatase activity. Supplied as a prepared solution, it is intended for use in cell-based and biochemical assays where exogenous ceramide is required.
  • Modulates ceramide signaling and apoptosis.
  • Activates serine/threonine protein phosphatases.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity for reproducible experimental results.
  • Suitable for cell-based and biochemical assays when diluted appropriately.