Learn More
Medchemexpress LLC C2 ceramide (ceramide 2) | 3102-57-6 | MFCD00153903 | 99.9% | 341.53 | C20H39NO3 | 1 MG
SDP
Catalog No. 5000212877
Encompass_Preferred
C2 Ceramide is a short-chain synthetic ceramide used as a research reagent and lipid signaling modulator. It acts as an activator of protein phosphatases and is commonly employed in studies of apoptosis, membrane biology, and cell-signaling pathways. Supplied as a solid for laboratory research use only.
  • Short-chain ceramide used in biochemical and cell-signaling research.
  • Activates protein phosphatases PP1 and PP2A, useful for phosphatase-related studies.
  • High purity (99.85%) suitable for research applications.
  • Soluble in ethanol and DMSO with ultrasonic warming as needed.
  • Available in small research-scale quantities for laboratory experiments.
Catalog No. 50-002-12877 Supplier Medchemexpress LLC Supplier No. HY1011801MG
C2 Ceramide is a short-chain synthetic ceramide used as a research reagent and lipid signaling modulator. It acts as an activator of protein phosphatases and is commonly employed in studies of apoptosis, membrane biology, and cell-signaling pathways. Supplied as a solid for laboratory research use only.
  • Short-chain ceramide used in biochemical and cell-signaling research.
  • Activates protein phosphatases PP1 and PP2A, useful for phosphatase-related studies.
  • High purity (99.85%) suitable for research applications.
  • Soluble in ethanol and DMSO with ultrasonic warming as needed.
  • Available in small research-scale quantities for laboratory experiments.