Learn More
Medchemexpress LLC C2 ceramide (ceramide 2) | 3102-57-6 | MFCD00153903 | 99.9% | 341.53 g/mol | C20H39NO3 | 10 MG
SDP
Catalog No. 5000211460
Encompass_Preferred
C2 ceramide (N-acetylsphingosine) is a short-chain ceramide used in biochemical and cell biology research as a lipid signaling molecule and protein phosphatase activator. It modulates cellular processes such as apoptosis, autophagy, and differentiation, and is applied in studies of mitochondrial function and epidermal barrier biology.
  • Activates protein phosphatases PP1 and PP2A.
  • Modulates apoptosis, autophagy, and cell differentiation.
  • Cell-permeable short-chain ceramide for signaling studies.
  • High reported purity for reproducible experiments.
  • Applicable to mitochondrial function and skin barrier assays.
Catalog No. 50-002-11460 Supplier Medchemexpress LLC Supplier No. HY10118010MG
C2 ceramide (N-acetylsphingosine) is a short-chain ceramide used in biochemical and cell biology research as a lipid signaling molecule and protein phosphatase activator. It modulates cellular processes such as apoptosis, autophagy, and differentiation, and is applied in studies of mitochondrial function and epidermal barrier biology.
  • Activates protein phosphatases PP1 and PP2A.
  • Modulates apoptosis, autophagy, and cell differentiation.
  • Cell-permeable short-chain ceramide for signaling studies.
  • High reported purity for reproducible experiments.
  • Applicable to mitochondrial function and skin barrier assays.