Learn More
Medchemexpress LLC C2 ceramide (N-acetylsphingosine) | 3102-57-6 | MFCD07374281 | 99.9% | 341.53 g/mol | C20H39NO3 | 25 MG
SDP
Catalog No. 5000211484
Encompass_Preferred
C2 ceramide (N-acetylsphingosine) is a short-chain ceramide research reagent used to modulate ceramide signaling and study cellular responses. It is used in biochemical and cell biology assays to probe protein phosphatase activity, mitochondrial function, and processes such as apoptosis, autophagy, and epidermal barrier behavior.
  • Activates protein phosphatases PP1 and PP2A for signaling studies.
  • Induces apoptosis and autophagy in cellular models.
  • Affects mitochondrial respiratory chain for metabolic research.
  • High-purity solid suitable for biochemical and cell-based assays.
  • Stable when stored sealed at -20°C; follow solvent storage guidelines for solutions.
  • Applicable to skin barrier and epidermal function research.
Catalog No. 50-002-11484 Supplier Medchemexpress LLC Supplier No. HY10118025MG
C2 ceramide (N-acetylsphingosine) is a short-chain ceramide research reagent used to modulate ceramide signaling and study cellular responses. It is used in biochemical and cell biology assays to probe protein phosphatase activity, mitochondrial function, and processes such as apoptosis, autophagy, and epidermal barrier behavior.
  • Activates protein phosphatases PP1 and PP2A for signaling studies.
  • Induces apoptosis and autophagy in cellular models.
  • Affects mitochondrial respiratory chain for metabolic research.
  • High-purity solid suitable for biochemical and cell-based assays.
  • Stable when stored sealed at -20°C; follow solvent storage guidelines for solutions.
  • Applicable to skin barrier and epidermal function research.