Learn More
Medchemexpress LLC C2/Complement C2 Hu 500ug
SDP
Catalog No. 5000178882
Encompass_Preferred
C2/Complement C2 protein is essential in the classical pathway of the complement system Activated by factor C1 C2 is cleaved into C2b and C2a As a serine protease C2a interacts with C4b generating the C3 or C5 convertase This crucial process activates and amplifies the complement cascade C2/Complement C2 Protein Human (HEK293 His) is the recombinant human-derived C2/Complement C2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78882 Supplier Medchemexpress LLC Supplier No. HYP74375500UG
C2/Complement C2 protein is essential in the classical pathway of the complement system Activated by factor C1 C2 is cleaved into C2b and C2a As a serine protease C2a interacts with C4b generating the C3 or C5 convertase This crucial process activates and amplifies the complement cascade C2/Complement C2 Protein Human (HEK293 His) is the recombinant human-derived C2/Complement C2 protein expressed by HEK293 with C-His labeled tag